missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylpyrimidine, 98%

Catalog No. p-7080682
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1036314 25 g
AAA1036322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1036314 Supplier Thermo Scientific Chemicals Supplier No. A1036314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5424-21-5 | C5H4Cl2N2 | 163.001 g/mol

2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.

Solubility
Soluble in Chloroform, Ether, Ethyl Acetate and Toluene. Insoluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5424-21-5
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00006064
InChI Key BTLKROSJMNFSQZ-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dichloro-6-methyl, 2,6-dichloro-4-methylpyrimidine, 2,4-dichloro-6-methyl-pyrimidine, 2,4-dichloro-6-methyl pyrimidine, 6-methyl-2,4-dichloropyrimidine, 2,4-dichlor-6-methylpyrimidin, pubchem6911, acmc-1b11m, ksc495i8r, 2,4-dichloro6-methylpyrimidine
PubChem CID 79471
IUPAC Name 2,4-dichloro-6-methylpyrimidine
SMILES CC1=CC(=NC(=N1)Cl)Cl

Specifications

Melting Point 44°C to 47°C
Boiling Point 219°C
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN1759
Beilstein 114295
Solubility Information Soluble in Chloroform,Ether,Ethyl Acetate and Toluene. Insoluble in water.
Formula Weight 163.01
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-6-methylpyrimidine
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 226-563-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only