missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylpyridine, 97+%

Catalog No. AAH6424806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6424806 5 g
AAH6424803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6424806 Supplier Thermo Scientific Chemicals Supplier No. H6424806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 42779-56-6 | C6H5Cl2N | 162.013 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 42779-56-6
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD08669834
InChI Key WUGTXQVNSRFDNV-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-picoline, 2,4-dichloro-6-methyl-pyridine, pubchem5465, d0ri6l, 2-methyl-4,6-dichloropyridine, pyridine,2,4-dichloro-6-methyl
PubChem CID 11959083
IUPAC Name 2,4-dichloro-6-methylpyridine
SMILES CC1=NC(=CC(=C1)Cl)Cl

Specifications

Boiling Point 205°C to 206°C
Quantity 5 g
Beilstein 111084
Solubility Information Slightly soluble in water.
Formula Weight 162.01
Percent Purity ≥97%
Chemical Name or Material 2,4-Dichloro-6-methylpyridine
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only