missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6,7-dimethoxyquinazoline 98.0+%, TCI America™

Catalog No. D37465G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37465G Supplier TCI America Supplier No. D37465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27631-29-4
Molecular Formula C10H8Cl2N2O2
Molecular Weight (g/mol) 259.086
MDL Number MFCD00051733
InChI Key DGHKCBSVAZXEPP-UHFFFAOYSA-N
Synonym 2,4-dichloro-6,7-dimethoxy-quinazoline, 2,4-dichloro-6,7-dimethoxy quinazoline, quinazoline, 2,4-dichloro-6,7-dimethoxy, 2-chloro-6,7-dimethoxyquinazolin-4-yl chloride, quinazoline,2,4-dichloro-6,7-dimethoxy, doxazosin impurity e, doxazosin mesilate impurity e ep, pubchem9430, usp doxazosin related compound e usp, acmc-209gyd
PubChem CID 520327
IUPAC Name 2,4-dichloro-6,7-dimethoxyquinazoline
SMILES COC1=C(C=C2C(=C1)C(=NC(=N2)Cl)Cl)OC

Specifications

Melting Point 175°C
Quantity 5 g
Formula Weight 259.09
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-6,7-dimethoxyquinazoline