missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6,7-dimethoxyquinazoline, 97%

CAS: 27631-29-4 | C10H8Cl2N2O2 | 259.086 g/mol

364,80 $ - 364,80 $

Identifiants chimiques

CAS 27631-29-4
MDL Number MFCD00051733
InChI Key DGHKCBSVAZXEPP-UHFFFAOYSA-N
PubChem CID 520327
IUPAC Name 2,4-dichloro-6,7-dimethoxyquinazoline
SMILES COC1=C(C=C2C(=C1)C(=NC(=N2)Cl)Cl)OC
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC342550250
FDS Afficher les documents
Thermo Scientific Chemicals
342550250
25 g Glass bottle N/A
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

27631-29-4
DGHKCBSVAZXEPP-UHFFFAOYSA-N
2,4-dichloro-6,7-dimethoxyquinazoline
MFCD00051733
520327
COC1=C(C=C2C(=C1)C(=NC(=N2)Cl)Cl)OC

Spécifications

27631-29-4
100.0
Beige to Yellow
97%
C10H8Cl2N2O2
25 g
DGHKCBSVAZXEPP-UHFFFAOYSA-N
2,4-dichloro-6,7-dimethoxyquinazoline
520327
≥96.0%
2, 4-Dichloro-6, 7-dimethoxyquinazoline, 97%
96.0
172°C to 177°C
Authentic
Glass bottle
MFCD00051733
2,4-dichloro-6,7-dimethoxy-quinazoline, 2,4-dichloro-6,7-dimethoxy quinazoline, quinazoline, 2,4-dichloro-6,7-dimethoxy, 2-chloro-6,7-dimethoxyquinazolin-4-yl chloride, quinazoline,2,4-dichloro-6,7-dimethoxy, doxazosin impurity e, doxazosin mesilate impurity e ep, pubchem9430, usp doxazosin related compound e usp, acmc-209gyd
COC1=C(C=C2C(=C1)C(=NC(=N2)Cl)Cl)OC
259.086
259.09
Crystalline Powder
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'tsca' : TSCA

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only