missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methoxypyrimidine 98.0+%, TCI America™

Catalog No. p-7127767
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35085G 5 g
D350825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35085G Supplier TCI America Supplier No. D35085G
Only null left

Chemical Identifiers

CAS 19646-07-2
Molecular Formula C5H4Cl2N2O
Molecular Weight (g/mol) 179.00
MDL Number MFCD02685670
InChI Key ZTHHRSBDBPCCMZ-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-methoxy-pyrimidine, 2,4-dichloro-5-methoxy pyrimidine, pyrimidine, 2,4-dichloro-5-methoxy, 2,4-dichloropyrimidin-5-yl methyl ether, 5-methoxy-2,4-dichloro pyrimidine, pubchem7076, ksc490s5l, 2,4 dichloro-5-methoxypyrimidine, 2,4-dichloro-5-methoxy-pyrimidin, 2,6-dichloro-5-methoxy pyrimidine
PubChem CID 601401
IUPAC Name 2,4-dichloro-5-methoxypyrimidine
SMILES COC1=CN=C(Cl)N=C1Cl

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 5 g
Formula Weight 179.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-5-methoxypyrimidine