missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methoxypyrimidine, 97%, Thermo Scientific Chemicals

Catalog No. p-7037423
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5615309 10 g
AAH5615322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5615309 Supplier Thermo Scientific Chemicals Supplier No. H5615309
Only null left

It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer’s disease.

Solubility
Slightly soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 19646-07-2
Molecular Formula C5H4Cl2N2O
Molecular Weight (g/mol) 179.00
MDL Number MFCD02685670
InChI Key ZTHHRSBDBPCCMZ-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-methoxy-pyrimidine, 2,4-dichloro-5-methoxy pyrimidine, pyrimidine, 2,4-dichloro-5-methoxy, 2,4-dichloropyrimidin-5-yl methyl ether, 5-methoxy-2,4-dichloro pyrimidine, pubchem7076, ksc490s5l, 2,4 dichloro-5-methoxypyrimidine, 2,4-dichloro-5-methoxy-pyrimidin, 2,6-dichloro-5-methoxy pyrimidine
PubChem CID 601401
IUPAC Name 2,4-dichloro-5-methoxypyrimidine
SMILES COC1=CN=C(Cl)N=C1Cl

Specifications

Melting Point 66°C to 70°C
Quantity 10 g
Solubility Information Slightly soluble in water.
Formula Weight 179
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-methoxypyrimidine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only