missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-fluoropyrimidine 98.0+%, TCI America™

$89.48 - $354.17

Chemical Identifiers

CAS 2927-71-1
Molecular Formula C4HCl2FN2
Molecular Weight (g/mol) 166.96
MDL Number MFCD00233551
InChI Key WHPFEQUEHBULBW-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-fluoro-pyrimidine, 5-fluoro-2,4-dichloropyrimidine, 2,6-dichloro-5-fluororacil, 2,4-dichloro-5-fluoro pyrimidine, pyrimidine, 2,4-dichloro-5-fluoro, 2,6-dichloro-5-fluoropyrimidine, 2,4-dichlor-5-fluorpyrimidin, pubchem5290, ksc204s2l, 2,4-dichloro-5fluoropyrimidine
PubChem CID 250705
IUPAC Name 2,4-dichloro-5-fluoropyrimidine
SMILES FC1=CN=C(Cl)N=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D33745G
View Documents
TCI America
D33745G
5 g
Each for $89.48
Only null left
 
D337425G
View Documents
TCI America
D337425G
25 g
Each for $354.17
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2927-71-1
166.96
WHPFEQUEHBULBW-UHFFFAOYSA-N
250705
FC1=CN=C(Cl)N=C1Cl
C4HCl2FN2
MFCD00233551
2,4-dichloro-5-fluoro-pyrimidine, 5-fluoro-2,4-dichloropyrimidine, 2,6-dichloro-5-fluororacil, 2,4-dichloro-5-fluoro pyrimidine, pyrimidine, 2,4-dichloro-5-fluoro, 2,6-dichloro-5-fluoropyrimidine, 2,4-dichlor-5-fluorpyrimidin, pubchem5290, ksc204s2l, 2,4-dichloro-5fluoropyrimidine
2,4-dichloro-5-fluoropyrimidine

Specifications

2927-71-1
80°C
MFCD00233551
2,4-dichloro-5-fluoro-pyrimidine, 5-fluoro-2,4-dichloropyrimidine, 2,6-dichloro-5-fluororacil, 2,4-dichloro-5-fluoro pyrimidine, pyrimidine, 2,4-dichloro-5-fluoro, 2,6-dichloro-5-fluoropyrimidine, 2,4-dichlor-5-fluorpyrimidin, pubchem5290, ksc204s2l, 2,4-dichloro-5fluoropyrimidine
FC1=CN=C(Cl)N=C1Cl
166.96
166.96
Crystalline Powder
39°C
C4HCl2FN2
5 g
WHPFEQUEHBULBW-UHFFFAOYSA-N
2,4-dichloro-5-fluoropyrimidine
250705
≥98.0% (GC)
2,4-Dichloro-5-fluoropyrimidine
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'