missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-1,3,5-triazine 98.0+%, TCI America™
$80.16 - $268.43
Chemical Identifiers
| CAS | 2831-66-5 |
|---|---|
| Molecular Formula | C3HCl2N3 |
| Molecular Weight (g/mol) | 149.962 |
| MDL Number | MFCD04115347 |
| InChI Key | OMRXVBREYFZQHU-UHFFFAOYSA-N |
| PubChem CID | 17810 |
| IUPAC Name | 2,4-dichloro-1,3,5-triazine |
| SMILES | C1=NC(=NC(=N1)Cl)Cl |
Chemical Identifiers
| 2831-66-5 | |
| 149.962 | |
| OMRXVBREYFZQHU-UHFFFAOYSA-N | |
| 2,4-dichloro-1,3,5-triazine |
| C3HCl2N3 | |
| MFCD04115347 | |
| 17810 | |
| C1=NC(=NC(=N1)Cl)Cl |
Specifications
| 2831-66-5 | |
| White | |
| C3HCl2N3 | |
| 1 g | |
| C1=NC(=NC(=N1)Cl)Cl | |
| 149.962 | |
| 149.96 | |
| Crystalline Powder |
| 56°C | |
| 102°C | |
| MFCD04115347 | |
| OMRXVBREYFZQHU-UHFFFAOYSA-N | |
| 2,4-dichloro-1,3,5-triazine | |
| 17810 | |
| ≥98.0% (GC,T) | |
| 2,4-Dichloro-1,3,5-triazine |
Safety and Handling
TSCA : No