missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-1-(2-iodophenoxy)benzene, ≥95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CD08469DA

 View more versions of this product

Catalog No. CD08469DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2, 4-Dichloro-1-(2-iodophenoxy)benzene
94.5
95%+
C12H7Cl2IO
MFCD00052444
2,4-dichloro-1-2-iodophenoxy benzene, 2,4-dichloro-1 2-iodophenoxy benzene, benzene,2,4-dichloro-1-2-iodophenoxy
ClC1=CC(Cl)=C(OC2=CC=CC=C2I)C=C1
364.99
364.99
175136-78-4
100.0
Amber Glass Bottle
Cl2C6H3OC6H4I
1g
ULAWXTPGHWKSDY-UHFFFAOYSA-N
2,4-dichloro-1-(2-iodophenoxy)benzene
2776994
95%

Chemical Identifiers

175136-78-4
364.99
ULAWXTPGHWKSDY-UHFFFAOYSA-N
2776994
ClC1=CC(Cl)=C(OC2=CC=CC=C2I)C=C1
C12H7Cl2IO
MFCD00052444
2,4-dichloro-1-2-iodophenoxy benzene, 2,4-dichloro-1 2-iodophenoxy benzene, benzene,2,4-dichloro-1-2-iodophenoxy
2,4-dichloro-1-(2-iodophenoxy)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions