missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dibutoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Supplier: TCI America D42631G
Specifications
| 2,4-Dibutoxyphenylboronic Acid (contains varying amounts of Anhydride) | |
| 104°C | |
| MFCD07784348 | |
| 2,4-Dibutoxybenzeneboronic Acid | |
| CCCCOC1=CC(OCCCC)=C(C=C1)B(O)O | |
| 266.14 | |
| 266.14 |
| 870778-89-5 | |
| C14H23BO4 | |
| 1 g | |
| RZGMDQYKUFKOMM-UHFFFAOYSA-N | |
| (2,4-dibutoxyphenyl)boronic acid | |
| 16217895 | |
| Crystalline Powder |
Chemical Identifiers
| 870778-89-5 | |
| 266.14 | |
| RZGMDQYKUFKOMM-UHFFFAOYSA-N | |
| 16217895 | |
| CCCCOC1=CC(OCCCC)=C(C=C1)B(O)O |
| C14H23BO4 | |
| MFCD07784348 | |
| 2,4-Dibutoxybenzeneboronic Acid | |
| (2,4-dibutoxyphenyl)boronic acid |
Safety and Handling
TSCA : No