missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromothiophene 90.0+%, TCI America™

Catalog No. D41461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41461G 1 g
D41465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41461G Supplier TCI America Supplier No. D41461G

Chemical Identifiers

CAS 3140-92-9
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.928
MDL Number MFCD00043889
InChI Key WAQFYSJKIRRXLP-UHFFFAOYSA-N
Synonym thiophene, 2,4-dibromo, 2,4 dibromothiophene, pubchem1908, 2,4-dibromo-thiophene, acmc-1cq64
PubChem CID 2724560
IUPAC Name 2,4-dibromothiophene
SMILES C1=C(SC=C1Br)Br

Specifications

Color Yellow
Boiling Point 97°C
Quantity 1 g
Formula Weight 241.93
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dibromothiophene
TSCA No
Recommended Storage Refrigerator