missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromothiazole 98.0+%, TCI America™

Catalog No. D36671G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36671G 1 g
D36675G 5 g
D366725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D36671G Supplier TCI America Supplier No. D36671G

Chemical Identifiers

CAS 4175-77-3
Molecular Formula C3HBr2NS
Molecular Weight (g/mol) 242.916
MDL Number MFCD01318994
InChI Key MKEJZKKVVUZXIS-UHFFFAOYSA-N
Synonym 2,4-dibromothiazole, 2,4-dibromo thiazole, dibromothiazole, 2.4-dibromothiazole, soxlllqacqdfwmijp@, 2,4-dibromo-thiazole, acmc-209jll, ksc235o5d, 2 pound not4-dibromo-thiazole
PubChem CID 2763297
IUPAC Name 2,4-dibromo-1,3-thiazole
SMILES C1=C(N=C(S1)Br)Br

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 1 g
Formula Weight 242.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromothiazole