missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromothiazole, 97%

Catalog No. AAH6023003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6023003 1 g
AAH6023006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6023003 Supplier Thermo Scientific Chemicals Supplier No. H6023003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4175-77-3 | C3HBr2NS | 242.916 g/mol

2,4-Dibromothiazole is applied as a building block in a synthesis of Melithiazole C employing a highly (E)-selective cross-metathesis between the derived 4-vinylthiazole and a dienote side-chain. It is also used as an intermediate in the preparation of various 2,4-disubstituted thiazole derivatives with biologically active properties such as survival motor neuron (SMN) protein modulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromothiazole is applied as a building block in a synthesis of Melithiazole C employing a highly (E)-selective cross-metathesis between the derived 4-vinylthiazole and a dienote side-chain. It is also used as an intermediate in the preparation of various 2,4-disubstituted thiazole derivatives with biologically active properties such as survival motor neuron (SMN) protein modulators.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 4175-77-3
Molecular Formula C3HBr2NS
Molecular Weight (g/mol) 242.916
MDL Number MFCD01318994
InChI Key MKEJZKKVVUZXIS-UHFFFAOYSA-N
Synonym 2,4-dibromothiazole, 2,4-dibromo thiazole, dibromothiazole, 2.4-dibromothiazole, soxlllqacqdfwmijp@, 2,4-dibromo-thiazole, acmc-209jll, ksc235o5d, 2 pound not4-dibromo-thiazole
PubChem CID 2763297
IUPAC Name 2,4-dibromo-1,3-thiazole
SMILES C1=C(N=C(S1)Br)Br

Specifications

Melting Point 80°C to 84°C
Quantity 1 g
Formula Weight 242.92
Percent Purity 97%
Chemical Name or Material 2,4-Dibromothiazole
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only