missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dibromopyridine 98.0+%, TCI America™
$152.86 - $507.02
Chemical Identifiers
| CAS | 58530-53-3 |
|---|---|
| Molecular Formula | C5H3Br2N |
| Molecular Weight (g/mol) | 236.89 |
| MDL Number | MFCD01859720 |
| InChI Key | PCMMSLVJMKQWMQ-UHFFFAOYSA-N |
| Synonym | 2,4-dibromo-pyridine, pyridine, 2,4-dibromo, 2,4-dibrompyridin, pubchem5452, acmc-1av5w, 2,4-dibromopyridine, ksc493e4t |
| PubChem CID | 817104 |
| IUPAC Name | 2,4-dibromopyridine |
| SMILES | BrC1=CC=NC(Br)=C1 |
Chemical Identifiers
| 58530-53-3 | |
| 236.89 | |
| PCMMSLVJMKQWMQ-UHFFFAOYSA-N | |
| 817104 | |
| BrC1=CC=NC(Br)=C1 |
| C5H3Br2N | |
| MFCD01859720 | |
| 2,4-dibromo-pyridine, pyridine, 2,4-dibromo, 2,4-dibrompyridin, pubchem5452, acmc-1av5w, 2,4-dibromopyridine, ksc493e4t | |
| 2,4-dibromopyridine |
Specifications
| 58530-53-3 | |
| White-Yellow | |
| C5H3Br2N | |
| 5 g | |
| PCMMSLVJMKQWMQ-UHFFFAOYSA-N | |
| 2,4-dibromopyridine | |
| 817104 | |
| ≥98.0% (GC) | |
| 2,4-Dibromopyridine |
| 40°C | |
| 238°C | |
| MFCD01859720 | |
| 2,4-dibromo-pyridine, pyridine, 2,4-dibromo, 2,4-dibrompyridin, pubchem5452, acmc-1av5w, 2,4-dibromopyridine, ksc493e4t | |
| BrC1=CC=NC(Br)=C1 | |
| 236.89 | |
| 236.89 | |
| Crystalline Powder |
Safety and Handling
TSCA : No