missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Diaminotoluene, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| Packaging | Ampule |
| CAS | 95-80-7 |
| Physical Form | Neat |
| Linear Formula | CH3C6H3(NH2)2 |
| MDL Number | MFCD00007804 |
| Melting Point | 97°C to 99°C |
| Boiling Point | 283°C to 285°C |
| Synonym | 4-Methyl-m-phenylenediamine; 2,4-Diaminotoluene; 2,4-Toluenediamine; 4-Methyl-1,3-phenylenediamine |
| Type | Organic Pollutant Standard |
| Quantity | 1000 mg |
| Show More |