Learn More
2,4-Diaminotoluene, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 95-80-7 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.17 |
| MDL Number | MFCD00007804 |
| InChI Key | VOZKAJLKRJDJLL-UHFFFAOYSA-N |
| Synonym | 2,4-diaminotoluene, 2,4-toluenediamine, developer t, toluene-2,4-diamine, 1,3-benzenediamine, 4-methyl, developer mtd, m-toluenediamine, fourrine m, m-tolylenediamine, benzofur mt |
| PubChem CID | 7261 |
| ChEBI | CHEBI:34237 |
| IUPAC Name | 4-methylbenzene-1,3-diamine |
| SMILES | CC1=CC=C(N)C=C1N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC146602500
|
Thermo Scientific Chemicals
146602500 |
250g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC146600010
|
Thermo Scientific Chemicals
146600010 |
1kg | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 95-80-7 | |
| 122.17 | |
| VOZKAJLKRJDJLL-UHFFFAOYSA-N | |
| 7261 | |
| 4-methylbenzene-1,3-diamine |
| C7H10N2 | |
| MFCD00007804 | |
| 2,4-diaminotoluene, 2,4-toluenediamine, developer t, toluene-2,4-diamine, 1,3-benzenediamine, 4-methyl, developer mtd, m-toluenediamine, fourrine m, m-tolylenediamine, benzofur mt | |
| CHEBI:34237 | |
| CC1=CC=C(N)C=C1N |
Specifications
| 95-80-7 | |
| 100.0 | |
| 149°C | |
| 97.5% min. (HPLC) | |
| C7H10N2 | |
| MFCD00007804 | |
| 13, 124 | |
| Solubility in water: 50g/L (25 c). Other solubilities: soluble in alcohol, ether, chloroform and, benzene | |
| CC1=CC=C(N)C=C1N | |
| 122.17 | |
| CHEBI:34237 | |
| 98% |
| 97.5 | |
| 283.0°C to 285.0°C | |
| Authentic | |
| Glass bottle | |
| CH3C6H3(NH2)2 | |
| 250g | |
| 2,4-diaminotoluene, 2,4-toluenediamine, developer t, toluene-2,4-diamine, 1,3-benzenediamine, 4-methyl, developer mtd, m-toluenediamine, fourrine m, m-tolylenediamine, benzofur mt | |
| VOZKAJLKRJDJLL-UHFFFAOYSA-N | |
| 4-methylbenzene-1,3-diamine | |
| 7261 | |
| 122.17 | |
| 2, 4-Diaminotoluene, 98% |
Safety and Handling
GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.
Toxic if swallowed.
Harmful in contact with skin.
May cause damage to organs through prolonged or repeated expos
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protectiv
GHS Signal Word: Danger
EINECSNumber : 202-453-1
RTECSNumber : XS9625000
TSCA : TSCA
RUO â Research Use Only