missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminotoluene, 98%

Catalog No. AAB2349530
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2349530 250 g
AAB234950B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2349530 Supplier Thermo Scientific Chemicals Supplier No. B2349530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-80-7 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-80-7
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007804
InChI Key VOZKAJLKRJDJLL-UHFFFAOYSA-N
Synonym 2,4-diaminotoluene, 2,4-toluenediamine, developer t, toluene-2,4-diamine, 1,3-benzenediamine, 4-methyl, developer mtd, m-toluenediamine, fourrine m, m-tolylenediamine, benzofur mt
PubChem CID 7261
ChEBI CHEBI:34237
IUPAC Name 4-methylbenzene-1,3-diamine
SMILES CC1=CC=C(N)C=C1N

Specifications

Melting Point 96°C to 101°C
Boiling Point 283°C to 285°C
Flash Point 169°C (336°F)
Odor Recognizable
Quantity 250 g
UN Number UN1709
Beilstein 2205839
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2,4-Diaminotoluene
Hazard Category H301-H312-H317-H341-H350-H361f-H373
Hazard Statement GHS H Statement
H301-H350-H341-H361-H373-H312-H317
Toxic if swallowed.
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P308+P313-P312-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 2,4-TOLUYLENEDIAMINE, SOLID
EINECSNumber 202-453-1
RTECSNumber XS9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only