missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Diaminodiphenylamine 98.0+%, TCI America™
$300.11 - $885.43
Chemical Identifiers
| CAS | 136-17-4 |
|---|---|
| Molecular Formula | C12H13N3 |
| Molecular Weight (g/mol) | 199.257 |
| MDL Number | MFCD00025285 |
| InChI Key | VOSLIUIVGWBSOK-UHFFFAOYSA-N |
| Synonym | 2,4-diaminodiphenylamine, diphenylamine, 2,4-diamino, n1-phenylbenzene-1,2,4-triamine, 1,2,4-benzenetriamine, n'-phenyl, unii-45dn363j8c, n sup 1-phenyl-1,2,4-benzenetriamine, 1,2,4-benzenetriamine, n sup 1-phenyl, diphenylamine,4-diamino, 1,4-benzenetriamine, n1-phenyl, 3-13-00-00552 beilstein handbook reference |
| PubChem CID | 8683 |
| IUPAC Name | 1-N-phenylbenzene-1,2,4-triamine |
| SMILES | C1=CC=C(C=C1)NC2=C(C=C(C=C2)N)N |
Chemical Identifiers
| 136-17-4 | |
| 199.257 | |
| VOSLIUIVGWBSOK-UHFFFAOYSA-N | |
| 8683 | |
| C1=CC=C(C=C1)NC2=C(C=C(C=C2)N)N |
| C12H13N3 | |
| MFCD00025285 | |
| 2,4-diaminodiphenylamine, diphenylamine, 2,4-diamino, n1-phenylbenzene-1,2,4-triamine, 1,2,4-benzenetriamine, n'-phenyl, unii-45dn363j8c, n sup 1-phenyl-1,2,4-benzenetriamine, 1,2,4-benzenetriamine, n sup 1-phenyl, diphenylamine,4-diamino, 1,4-benzenetriamine, n1-phenyl, 3-13-00-00552 beilstein handbook reference | |
| 1-N-phenylbenzene-1,2,4-triamine |
Specifications
| 136-17-4 | |
| Purple-White | |
| MFCD00025285 | |
| 2,4-diaminodiphenylamine, diphenylamine, 2,4-diamino, n1-phenylbenzene-1,2,4-triamine, 1,2,4-benzenetriamine, n'-phenyl, unii-45dn363j8c, n sup 1-phenyl-1,2,4-benzenetriamine, 1,2,4-benzenetriamine, n sup 1-phenyl, diphenylamine,4-diamino, 1,4-benzenetriamine, n1-phenyl, 3-13-00-00552 beilstein handbook reference | |
| C1=CC=C(C=C1)NC2=C(C=C(C=C2)N)N | |
| 199.257 | |
| 199.26 | |
| Crystalline Powder |
| 125°C | |
| C12H13N3 | |
| 5 g | |
| VOSLIUIVGWBSOK-UHFFFAOYSA-N | |
| 1-N-phenylbenzene-1,2,4-triamine | |
| 8683 | |
| ≥98.0% (T) | |
| 2,4-Diaminodiphenylamine |
Safety and Handling
RTECSNumber : DC1954000
TSCA : No