missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminodiphenylamine 98.0+%, TCI America™

Catalog No. D00875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00875G 5 g
D008725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00875G Supplier TCI America Supplier No. D00875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 136-17-4
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.257
MDL Number MFCD00025285
InChI Key VOSLIUIVGWBSOK-UHFFFAOYSA-N
Synonym 2,4-diaminodiphenylamine, diphenylamine, 2,4-diamino, n1-phenylbenzene-1,2,4-triamine, 1,2,4-benzenetriamine, n'-phenyl, unii-45dn363j8c, n sup 1-phenyl-1,2,4-benzenetriamine, 1,2,4-benzenetriamine, n sup 1-phenyl, diphenylamine,4-diamino, 1,4-benzenetriamine, n1-phenyl, 3-13-00-00552 beilstein handbook reference
PubChem CID 8683
IUPAC Name 1-N-phenylbenzene-1,2,4-triamine
SMILES C1=CC=C(C=C1)NC2=C(C=C(C=C2)N)N

Specifications

Melting Point 125°C
Color Purple-White
Quantity 5 g
Formula Weight 199.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diaminodiphenylamine
RTECSNumber DC1954000
TSCA No