Learn More
2,4-Diaminodiphenylamine, 98%
CAS: 136-17-4 | C12H13N3 | 199.257 g/mol
$192.42 - $748.49
Chemical Identifiers
| CAS | 136-17-4 |
|---|---|
| Molecular Formula | C12H13N3 |
| Molecular Weight (g/mol) | 199.257 |
| MDL Number | MFCD00025285 |
| InChI Key | VOSLIUIVGWBSOK-UHFFFAOYSA-N |
| Synonym | 2,4-diaminodiphenylamine, diphenylamine, 2,4-diamino, n1-phenylbenzene-1,2,4-triamine, 1,2,4-benzenetriamine, n'-phenyl, unii-45dn363j8c, n sup 1-phenyl-1,2,4-benzenetriamine, 1,2,4-benzenetriamine, n sup 1-phenyl, diphenylamine,4-diamino, 1,4-benzenetriamine, n1-phenyl, 3-13-00-00552 beilstein handbook reference |
| PubChem CID | 8683 |
| IUPAC Name | 1-N-phenylbenzene-1,2,4-triamine |
| SMILES | C1=CC=C(C=C1)NC2=C(C=C(C=C2)N)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2073503
|
Thermo Scientific Chemicals
B2073503 |
1 g |
Each for $192.42
|
|
|||||
|
AAB2073506
|
Thermo Scientific Chemicals
B2073506 |
5 g |
Each for $748.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 136-17-4 | |
| 199.257 | |
| VOSLIUIVGWBSOK-UHFFFAOYSA-N | |
| 8683 | |
| C1=CC=C(C=C1)NC2=C(C=C(C=C2)N)N |
| C12H13N3 | |
| MFCD00025285 | |
| 2,4-diaminodiphenylamine, diphenylamine, 2,4-diamino, n1-phenylbenzene-1,2,4-triamine, 1,2,4-benzenetriamine, n'-phenyl, unii-45dn363j8c, n sup 1-phenyl-1,2,4-benzenetriamine, 1,2,4-benzenetriamine, n sup 1-phenyl, diphenylamine,4-diamino, 1,4-benzenetriamine, n1-phenyl, 3-13-00-00552 beilstein handbook reference | |
| 1-N-phenylbenzene-1,2,4-triamine |
Specifications
| 136-17-4 | |
| C12H13N3 | |
| 1 g | |
| 2,4-diaminodiphenylamine, diphenylamine, 2,4-diamino, n1-phenylbenzene-1,2,4-triamine, 1,2,4-benzenetriamine, n'-phenyl, unii-45dn363j8c, n sup 1-phenyl-1,2,4-benzenetriamine, 1,2,4-benzenetriamine, n sup 1-phenyl, diphenylamine,4-diamino, 1,4-benzenetriamine, n1-phenyl, 3-13-00-00552 beilstein handbook reference | |
| C1=CC=C(C=C1)NC2=C(C=C(C=C2)N)N | |
| 199.257 | |
| 199.26 | |
| 2,4-Diaminodiphenylamine |
| 129°C to 131°C | |
| MFCD00025285 | |
| UN2811 | |
| VOSLIUIVGWBSOK-UHFFFAOYSA-N | |
| 1-N-phenylbenzene-1,2,4-triamine | |
| 8683 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c
H301+H311-H330-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only