missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminodiphenylamine, 98%

Catalog No. AAB2073503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2073503 1 g
AAB2073506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2073503 Supplier Thermo Scientific Chemicals Supplier No. B2073503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 136-17-4 | C12H13N3 | 199.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136-17-4
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.257
MDL Number MFCD00025285
InChI Key VOSLIUIVGWBSOK-UHFFFAOYSA-N
Synonym 2,4-diaminodiphenylamine, diphenylamine, 2,4-diamino, n1-phenylbenzene-1,2,4-triamine, 1,2,4-benzenetriamine, n'-phenyl, unii-45dn363j8c, n sup 1-phenyl-1,2,4-benzenetriamine, 1,2,4-benzenetriamine, n sup 1-phenyl, diphenylamine,4-diamino, 1,4-benzenetriamine, n1-phenyl, 3-13-00-00552 beilstein handbook reference
PubChem CID 8683
IUPAC Name 1-N-phenylbenzene-1,2,4-triamine
SMILES C1=CC=C(C=C1)NC2=C(C=C(C=C2)N)N

Specifications

Melting Point 129°C to 131°C
Quantity 1 g
UN Number UN2811
Formula Weight 199.26
Percent Purity 98%
Chemical Name or Material 2,4-Diaminodiphenylamine
Hazard Category H301+H311-H330-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only