missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-nitroquinazoline, 98%

Catalog No. AAL1233003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1233003 1 g
AAL1233006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1233003 Supplier Thermo Scientific Chemicals Supplier No. L1233003

CAS: 7154-34-9 | C8H7N5O2 | 205.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7154-34-9
Molecular Formula C8H7N5O2
Molecular Weight (g/mol) 205.18
MDL Number MFCD00023910
InChI Key YZMJNZRTRWPJFY-UHFFFAOYSA-N
Synonym 2,4-diamino-6-nitroquinazoline, 2,4-quinazolinediamine,6-nitro, 6-nitro-2,4-quinazolinediamine #, 2,4-diamino-6-nitro-quinazoline, 1a
PubChem CID 252163
IUPAC Name 6-nitroquinazoline-2,4-diamine
SMILES NC1=NC(N)=C2C=C(C=CC2=N1)[N+]([O-])=O

Specifications

Melting Point >360°C
Quantity 1 g
Beilstein 749531
Formula Weight 205.18
Percent Purity 98%
Chemical Name or Material 2,4-Diamino-6-nitroquinazoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only