missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-methyl-1,3,5-triazine 98.0+%, TCI America™

Catalog No. D058325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D058325G 25 g
D0583500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D058325G Supplier TCI America Supplier No. D058325G
Only null left

Chemical Identifiers

CAS 542-02-9
Molecular Formula C4H7N5
Molecular Weight (g/mol) 125.135
MDL Number MFCD00023192
InChI Key NJYZCEFQAIUHSD-UHFFFAOYSA-N
Synonym acetoguanamine, 2,4-diamino-6-methyl-1,3,5-triazine, 1,3,5-triazine-2,4-diamine, 6-methyl, 2-methyl-4,6-diamino-1,3,5-triazine, 2,4-diamino-6-methyl-s-triazine, 6-methyl-1,3,5-triazine-2,4-diyldiamine, s-triazine, 2,4-diamino-6-methyl, acetoguanamin, ccris 8909, dsstox_cid_9186
PubChem CID 10949
ChEBI CHEBI:72475
IUPAC Name 6-methyl-1,3,5-triazine-2,4-diamine
SMILES CC1=NC(=NC(=N1)N)N

Specifications

Color White
Quantity 25 g
Formula Weight 125.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-methyl-1,3,5-triazine
EINECSNumber (5)-1026
TSCA Yes