missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diacetylphloroglucinol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 452800010

 View more versions of this product

Catalog No. AC452800010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2,4-Diacetylphloroglucinol
Authentic
Glass Bottle
MFCD00100483
2,4-diacetylphloroglucinol, 1-3-acetyl-2,4,6-trihydroxyphenyl ethan-1-one, diacetylphloroglucinol, phloroglucinol, diacetyl, unii-8xv4yyo3wn, 8xv4yyo3wn, 1-3-acetyl-2,4,6-trihydroxyphenyl ethanone, 1,1'-2,4,6-trihydroxy-1,3-phenylene bisethanone, ethanone,1,1'-2,4,6-trihydroxy-1,3-phenylene bis, 1-3-acetyl-2,4,6-trihydroxy-phenyl ethanone
CC(=O)C1=C(O)C(C(C)=O)=C(O)C=C1O
210.19
CHEBI:78688
97%
2161-86-6
96% min. (HPLC)
C10H10O5
1g
PIFFQYJYNWXNGE-UHFFFAOYSA-N
1-(3-acetyl-2,4,6-trihydroxyphenyl)ethanone
16547
210.18

Chemical Identifiers

2161-86-6
210.19
PIFFQYJYNWXNGE-UHFFFAOYSA-N
16547
1-(3-acetyl-2,4,6-trihydroxyphenyl)ethanone
C10H10O5
MFCD00100483
2,4-diacetylphloroglucinol, 1-3-acetyl-2,4,6-trihydroxyphenyl ethan-1-one, diacetylphloroglucinol, phloroglucinol, diacetyl, unii-8xv4yyo3wn, 8xv4yyo3wn, 1-3-acetyl-2,4,6-trihydroxyphenyl ethanone, 1,1'-2,4,6-trihydroxy-1,3-phenylene bisethanone, ethanone,1,1'-2,4,6-trihydroxy-1,3-phenylene bis, 1-3-acetyl-2,4,6-trihydroxy-phenyl ethanone
CHEBI:78688
CC(=O)C1=C(O)C(C(C)=O)=C(O)C=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only