missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)indole, 98%

Catalog No. AAA1363803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1363803 1 g
AAA1363806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1363803 Supplier Thermo Scientific Chemicals Supplier No. A1363803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1211-35-4 | C14H10ClN | 227.691 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1211-35-4
Molecular Formula C14H10ClN
Molecular Weight (g/mol) 227.691
MDL Number MFCD00047159
InChI Key KDNXKQSAAZNUCK-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl-1h-indole, 2-4-chlorophenyl indole, indole, 2-4-chlorophenyl, 2-p-chlorophenyl indole, maybridge1_005860, 2-p-chlorphenyl-indol, 2-4-chlorophenyl-indole, 2-4-chlorphenyl-1h-indole, 1h-indole,2-4-chlorophenyl, 2-4-chloro-phenyl-1h-indole
PubChem CID 220463
IUPAC Name 2-(4-chlorophenyl)-1H-indole
SMILES C1=CC=C2C(=C1)C=C(N2)C3=CC=C(C=C3)Cl

Specifications

Melting Point 205°C to 207°C
Quantity 1 g
Formula Weight 227.7
Percent Purity 98%
Chemical Name or Material 2-(4-Chlorophenyl)indole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only