missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)ethanol 98.0+%, TCI America™

Catalog No. p-7125347
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C22335G 5 g
C223325G 25 g
2 options

Catalog No. C22335G

Supplier: TCI America C22335G

Only null left

Chemical Identifiers

CAS 1875-88-3
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00002899
InChI Key HZFRKZWBVUJYDA-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl ethanol, 4-chlorophenethylalcohol, 4-chlorophenethyl alcohol, 2-4-chlorophenyl ethan-1-ol, benzeneethanol, 4-chloro, p-chlorophenethylic alcohol, 4-chlorophenethanol, 4-chlorophenyl methyl carbinol, 4-chlorobenzeneethanol, p-chlorophenethylalcohol
PubChem CID 74647
IUPAC Name 2-(4-chlorophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)Cl

Specifications

Color Yellow
Boiling Point 153°C
Quantity 5 g
Physical Form Clear Liquid at 20°C
Formula Weight 156.61
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-(4-Chlorophenyl)ethanol