missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)-3-methylbutyric Acid 98.0+%, TCI America™

Catalog No. C274125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C274125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C274125G Supplier TCI America Supplier No. C274125G
Only null left

Chemical Identifiers

CAS 2012-74-0
Molecular Formula C11H13ClO2
Molecular Weight (g/mol) 212.67
MDL Number MFCD04112625,MFCD11858304,MFCD00037763
InChI Key VTJMSIIXXKNIDJ-UHFFFAOYNA-N
PubChem CID 16197
ChEBI CHEBI:39345
IUPAC Name 2-(4-chlorophenyl)-3-methylbutanoic acid
SMILES CC(C)C(C(O)=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 25 g
Formula Weight 212.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Chlorophenyl)-3-methylbutyric Acid
EINECSNumber (3)-3472
RTECSNumber ES8450000
TSCA Yes