missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(chloromethyl)piperidino]pyrimidine, Thermo Scientific™

Chemical Identifiers

CAS 111247-62-2
Molecular Formula C10H14ClN3
Molecular Weight (g/mol) 211.693
InChI Key XEOONRMTEFSPSW-UHFFFAOYSA-N
Synonym 2-4-chloromethyl piperidin-1-yl pyrimidine, 2-4-chloromethyl piperidino pyrimidine, 2-4-chloromethyl-piperidin-1-yl-pyrimidine, 2-4-chloromethyl piperidyl pyrimidine, 2-4-chloromethylpiperidin-1-yl pyrimidine
PubChem CID 14554091
IUPAC Name 2-[4-(chloromethyl)piperidin-1-yl]pyrimidine
SMILES C1CN(CCC1CCl)C2=NC=CC=N2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC20724CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC20724DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC20724DE
View Documents Promotion Details
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

111247-62-2
211.693
2-4-chloromethyl piperidin-1-yl pyrimidine, 2-4-chloromethyl piperidino pyrimidine, 2-4-chloromethyl-piperidin-1-yl-pyrimidine, 2-4-chloromethyl piperidyl pyrimidine, 2-4-chloromethylpiperidin-1-yl pyrimidine
2-[4-(chloromethyl)piperidin-1-yl]pyrimidine
C10H14ClN3
XEOONRMTEFSPSW-UHFFFAOYSA-N
14554091
C1CN(CCC1CCl)C2=NC=CC=N2

Specifications

111247-62-2
2-4-chloromethyl piperidin-1-yl pyrimidine, 2-4-chloromethyl piperidino pyrimidine, 2-4-chloromethyl-piperidin-1-yl-pyrimidine, 2-4-chloromethyl piperidyl pyrimidine, 2-4-chloromethylpiperidin-1-yl pyrimidine
C1CN(CCC1CCl)C2=NC=CC=N2
211.693
211.69
2-[4-(chloromethyl)piperidino]pyrimidine
C10H14ClN3
XEOONRMTEFSPSW-UHFFFAOYSA-N
2-[4-(chloromethyl)piperidin-1-yl]pyrimidine
14554091
97%
Videos
SDS
Documents

Documents