missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Chloromethyl)phenyl]pyrimidine, 97%, Thermo Scientific™

Catalog No. p-3541010
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC56224CB 250mg Amber Glass Bottle
CC56224DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC56224CB Supplier Thermo Scientific Chemicals Supplier No. CC56224CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 898289-48-0
Molecular Formula C11H9ClN2
Molecular Weight (g/mol) 204.657
MDL Number MFCD09702381
InChI Key ZYZLFVLHAHBXGB-UHFFFAOYSA-N
Synonym 2-4-chloromethyl phenyl pyrimidine, 4-pyrimidin-2-ylbenzyl chloride, 4-pyrimidin-2-yl benzyl chloride
PubChem CID 24229560
IUPAC Name 2-[4-(chloromethyl)phenyl]pyrimidine
SMILES C1=CN=C(N=C1)C2=CC=C(C=C2)CCl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 204.66
Percent Purity 97%
Chemical Name or Material 2-[4-(Chloromethyl)phenyl]pyrimidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.