missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorobenzoyl)pyridine 97.0+%, TCI America™

Catalog No. C26475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26475G 5 g
C264725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26475G Supplier TCI America Supplier No. C26475G
Only null left

Chemical Identifiers

CAS 6318-51-0
Molecular Formula C12H8ClNO
Molecular Weight (g/mol) 217.65
MDL Number MFCD02930888
InChI Key KHXSJSBQIWAIEG-UHFFFAOYSA-N
Synonym 4-Chlorophenyl 2-Pyridyl Ketone
PubChem CID 80594
IUPAC Name 2-(4-chlorobenzoyl)pyridine
SMILES ClC1=CC=C(C=C1)C(=O)C1=CC=CC=N1

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 160°C
Quantity 5 g
Formula Weight 217.65
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Chlorobenzoyl)pyridine