missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)pyridine 98.0+%, TCI America™

Catalog No. p-7123580
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45001G 1 g
B45005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45001G Supplier TCI America Supplier No. B45001G

Chemical Identifiers

CAS 63996-36-1
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD04116231
InChI Key FBQFCXDBCPREBP-UHFFFAOYSA-N
PubChem CID 1515237
IUPAC Name 2-(4-bromophenyl)pyridine
SMILES C1=CC=NC(=C1)C2=CC=C(C=C2)Br

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 1 g
Formula Weight 234.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)pyridine