missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)ethylamine 98.0+%, TCI America™

Catalog No. B34545G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34545G 5 g
B34541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34545G Supplier TCI America Supplier No. B34545G

Chemical Identifiers

CAS 73918-56-6
Molecular Formula C8H11BrN
Molecular Weight (g/mol) 201.09
MDL Number MFCD00008189
InChI Key ZSZCXAOQVBEPME-UHFFFAOYSA-O
Synonym 4-bromophenethylamine, 2-4-bromophenyl ethylamine, 2-4-bromophenyl ethanamine, p-bromophenethylamine, 2-4-bromo-phenyl-ethylamine, benzeneethanamine, 4-bromo, 2-4-bromophenyl ethan-1-amine, 4-bromophenylethylamine, 4-bromobenzeneethanamine, 1-amino-2-4-bromophenyl ethane
PubChem CID 533915
IUPAC Name 2-(4-bromophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=C(Br)C=C1

Specifications

Color Yellow
Boiling Point 72°C
Quantity 5 g
UN Number 2735
Formula Weight 200.08
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(4-Bromophenyl)ethylamine