missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)benzimidazole 98.0+%, TCI America™

Catalog No. B28121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28121G 1 g
B28125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28121G Supplier TCI America Supplier No. B28121G

Chemical Identifiers

CAS 2622-74-4
Molecular Formula C13H9BrN2
Molecular Weight (g/mol) 273.13
MDL Number MFCD00224360
InChI Key YRWMGOSKROWAIT-UHFFFAOYSA-N
Synonym 2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1h-benzo d imidazole, 2-4-bromophenyl-1h-benzimidazole, 2-4-bromophenyl-1h-benzoimidazole, 2-4-bromophenyl benzoimidazole, 2-4-bromo-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-4-bromophenyl, 2-4-bromophenyl-1h-1,3-benzodiazole, acmc-209gpy, boron-mol bm885
PubChem CID 286182
IUPAC Name 2-(4-bromophenyl)-1H-1,3-benzodiazole
SMILES BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1

Specifications

Melting Point 299°C
Quantity 1 g
Formula Weight 273.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)benzimidazole