missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromobutoxy)tetrahydro-2H-pyran 98.0+%, TCI America™

Catalog No. B31865G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B31865G Supplier TCI America Supplier No. B31865G
Only null left

Chemical Identifiers

CAS 31608-22-7
Molecular Formula C9H17BrO2
Molecular Weight (g/mol) 237.14
MDL Number MFCD06654117
InChI Key DFXFQWZHKKEJDM-UHFFFAOYNA-N
PubChem CID 559019
IUPAC Name 2-(4-bromobutoxy)oxane
SMILES BrCCCCOC1CCCCO1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 237.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Bromobutoxy)tetrahydro-2H-pyran
TSCA No
Recommended Storage Freezer