missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Bis[4-(diethylamino)-2-hydroxyphenyl]squaraine 98.0+%, TCI America™
Supplier: TCI America B43421G
Specifications
| 2,4-Bis[4-(diethylamino)-2-hydroxyphenyl]squaraine | |
| C24H28N2O4 | |
| 1 g | |
| CCN(CC)C1=CC(=C(C=C1)C2=C(C(=C3C=CC(=CC3=O)N(CC)CC)C2=O)O)O | |
| 408.498 | |
| 408.50 | |
| Crystalline Powder |
| 68842-66-0 | |
| MFCD00142492 | |
| ODWFNEITXILTAS-RELWKKBWSA-N | |
| (6E)-3-(diethylamino)-6-[3-[4-(diethylamino)-2-hydroxyphenyl]-2-hydroxy-4-oxocyclobut-2-en-1-ylidene]cyclohexa-2,4-dien-1-one | |
| 18806793 | |
| ≥98.0% (N) |
Chemical Identifiers
| 68842-66-0 | |
| 408.498 | |
| ODWFNEITXILTAS-RELWKKBWSA-N | |
| (6E)-3-(diethylamino)-6-[3-[4-(diethylamino)-2-hydroxyphenyl]-2-hydroxy-4-oxocyclobut-2-en-1-ylidene]cyclohexa-2,4-dien-1-one |
| C24H28N2O4 | |
| MFCD00142492 | |
| 18806793 | |
| CCN(CC)C1=CC(=C(C=C1)C2=C(C(=C3C=CC(=CC3=O)N(CC)CC)C2=O)O)O |
Safety and Handling
TSCA : No