missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine 98.0+%, TCI America™
$203.17 - $706.48
Chemical Identifiers
| CAS | 182918-13-4 |
|---|---|
| Molecular Formula | C27H18ClN3 |
| Molecular Weight (g/mol) | 419.912 |
| MDL Number | MFCD27979765 |
| InChI Key | FVQBRDRAILXTMJ-UHFFFAOYSA-N |
| PubChem CID | 22476006 |
| IUPAC Name | 2-chloro-4,6-bis(4-phenylphenyl)-1,3,5-triazine |
| SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)C3=NC(=NC(=N3)Cl)C4=CC=C(C=C4)C5=CC=CC=C5 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4941200MG
|
TCI America
B4941200MG |
200 mg |
Each for $203.17
|
|
|||||
|
B49411G
|
TCI America
B49411G |
1 g |
Each for $706.48
|
|
|||||
Chemical Identifiers
| 182918-13-4 | |
| 419.912 | |
| FVQBRDRAILXTMJ-UHFFFAOYSA-N | |
| 2-chloro-4,6-bis(4-phenylphenyl)-1,3,5-triazine |
| C27H18ClN3 | |
| MFCD27979765 | |
| 22476006 | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)C3=NC(=NC(=N3)Cl)C4=CC=C(C=C4)C5=CC=CC=C5 |
Specifications
| 182918-13-4 | |
| White-Yellow | |
| MFCD27979765 | |
| FVQBRDRAILXTMJ-UHFFFAOYSA-N | |
| 2-chloro-4,6-bis(4-phenylphenyl)-1,3,5-triazine | |
| 22476006 | |
| ≥98.0% (HPLC,N) | |
| 2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine |
| 217°C | |
| C27H18ClN3 | |
| 200 mg | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)C3=NC(=NC(=N3)Cl)C4=CC=C(C=C4)C5=CC=CC=C5 | |
| 419.912 | |
| 419.91 | |
| Crystalline Powder |
Safety and Handling
TSCA : No