missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine 97.0+%, TCI America™
$227.41 - $667.33
Chemical Identifiers
| CAS | 2725-22-6 |
|---|---|
| Molecular Formula | C33H39N3O2 |
| Molecular Weight (g/mol) | 509.69 |
| MDL Number | MFCD09834146 |
| InChI Key | DUSCWEXFMYJRHL-UHFFFAOYSA-N |
| Synonym | 2-[4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl]-5-(n-octyloxy)phenol |
| PubChem CID | 6850826 |
| IUPAC Name | 6-[4,6-bis(2,4-dimethylphenyl)-2,5-dihydro-1,3,5-triazin-2-ylidene]-2-(octyloxy)cyclohexa-2,4-dien-1-one |
| SMILES | CCCCCCCCOC1=CC=CC(C1=O)=C1N=C(NC(=N1)C1=CC=C(C)C=C1C)C1=CC=C(C)C=C1C |
Chemical Identifiers
| 2725-22-6 | |
| 509.69 | |
| DUSCWEXFMYJRHL-UHFFFAOYSA-N | |
| 6850826 | |
| CCCCCCCCOC1=CC=CC(C1=O)=C1N=C(NC(=N1)C1=CC=C(C)C=C1C)C1=CC=C(C)C=C1C |
| C33H39N3O2 | |
| MFCD09834146 | |
| 2-[4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl]-5-(n-octyloxy)phenol | |
| 6-[4,6-bis(2,4-dimethylphenyl)-2,5-dihydro-1,3,5-triazin-2-ylidene]-2-(octyloxy)cyclohexa-2,4-dien-1-one |
Specifications
| 2725-22-6 | |
| Yellow | |
| C33H39N3O2 | |
| 5 g | |
| DUSCWEXFMYJRHL-UHFFFAOYSA-N | |
| 6-[4,6-bis(2,4-dimethylphenyl)-2,5-dihydro-1,3,5-triazin-2-ylidene]-2-(octyloxy)cyclohexa-2,4-dien-1-one | |
| 6850826 | |
| ≥97.0% (N) | |
| 2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine |
| 90°C | |
| 695°C | |
| MFCD09834146 | |
| 2-[4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl]-5-(n-octyloxy)phenol | |
| CCCCCCCCOC1=CC=CC(C1=O)=C1N=C(NC(=N1)C1=CC=C(C)C=C1C)C1=CC=C(C)C=C1C | |
| 509.69 | |
| 509.69 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6335
TSCA : Yes