missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine 97.0+%, TCI America™

Catalog No. b4940200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4940200MG 200 mg
B49401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4940200MG Supplier TCI America Supplier No. B4940200MG
Only null left

Chemical Identifiers

CAS 10202-45-6
Molecular Formula C15H9Cl2N3
Molecular Weight (g/mol) 302.158
InChI Key JYPGHMDTTDKUEL-UHFFFAOYSA-N
PubChem CID 22352243
IUPAC Name 2,4-dichloro-6-(4-phenylphenyl)-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=NC(=NC(=N3)Cl)Cl

Specifications

Melting Point 159°C
Color Yellow
Quantity 200 mg
Formula Weight 302.16
Percent Purity ≥97.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine