missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Biphenylyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. B30195G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B30195G Supplier TCI America Supplier No. B30195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144432-80-4
Molecular Formula C18H21BO2
Molecular Weight (g/mol) 280.17
MDL Number MFCD07368291
InChI Key REDKQKNJWVIPIO-UHFFFAOYSA-N
Synonym 4-biphenylboronic acid pinacol ester, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-biphenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-phenylphenyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, 2-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl diphenyl
PubChem CID 11989651
IUPAC Name 2-{[1,1'-biphenyl]-4-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 5 g
Formula Weight 280.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Biphenylyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane