missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Aminophenyl)ethylamine 97.0+%, TCI America™

Catalog No. A15095G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15095G 5 g
A15091G 1 g
A150925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A15095G Supplier TCI America Supplier No. A15095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13472-00-9
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00008194
InChI Key LNPMZQXEPNWCMG-UHFFFAOYSA-N
Synonym 2-4-aminophenyl ethylamine, 4-2-aminoethyl aniline, 4-aminophenethylamine, 4-2-amino-ethyl-phenylamine, p-aminophenethylamine, benzeneethanamine, 4-amino, 4-2-aminoethyl benzenamine, 4-aminophenylethylamine, 4-2-aminoethyl phenylamine, 4-2-amino-ethyl-aniline
PubChem CID 72895
IUPAC Name 4-(2-aminoethyl)aniline
SMILES C1=CC(=CC=C1CCN)N

Specifications

Boiling Point 142°C
Quantity 5 g
UN Number 3259
Formula Weight 136.20
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-(4-Aminophenyl)ethylamine
EINECSNumber (3)-0331
TSCA No