missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Aminophenyl)ethylamine, 98%

Catalog No. AAL1578203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1578203 1 g
AAL1578206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1578203 Supplier Thermo Scientific Chemicals Supplier No. L1578203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13472-00-9 | C8H12N2 | 136.198 g/mol

4-(2-Aminoethyl)aniline has been used in chemical modification of silk fibroin to tailor the overall hydrophilicity and structure of silk. It has been used as reagent in polycondensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Aminoethyl)aniline has been used in chemical modification of silk fibroin to tailor the overall hydrophilicity and structure of silk. It has been used as reagent in polycondensation reactions.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 13472-00-9
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00008194
InChI Key LNPMZQXEPNWCMG-UHFFFAOYSA-N
Synonym 2-4-aminophenyl ethylamine, 4-2-aminoethyl aniline, 4-aminophenethylamine, 4-2-amino-ethyl-phenylamine, p-aminophenethylamine, benzeneethanamine, 4-amino, 4-2-aminoethyl benzenamine, 4-aminophenylethylamine, 4-2-aminoethyl phenylamine, 4-2-amino-ethyl-aniline
PubChem CID 72895
IUPAC Name 4-(2-aminoethyl)aniline
SMILES C1=CC(=CC=C1CCN)N

Specifications

Melting Point 28°C to 31°C
Density 1.057
Boiling Point 103°C (0.3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.591
Quantity 1 g
UN Number UN3259
Beilstein 1099913
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 2-(4-Aminophenyl)ethylamine
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 236-739-2
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only