missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(trifluoromethyl)-1,3,5-triazine, 98%

Catalog No. p-7026162
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2200103 1 g
AAB2200106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2200103 Supplier Thermo Scientific Chemicals Supplier No. B2200103
Only null left

CAS: 368-66-1 | C6F9N3 | 285.07 g/mol

2,4,6-Tris(trifluoromethyl)-1,3,5-triazine is used to prepare 1-phenyl-4,6-bis-trifluoromethyl-1H-pyrazolo[3,4-d]pyrimidine by reacting with sodium 5-amino-1-phenyl-1H-pyrazole-4-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine is used to prepare 1-phenyl-4,6-bis-trifluoromethyl-1H-pyrazolo[3,4-d]pyrimidine by reacting with sodium 5-amino-1-phenyl-1H-pyrazole-4-carboxylate.

Notes
Air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 368-66-1
Molecular Formula C6F9N3
Molecular Weight (g/mol) 285.07
MDL Number MFCD00042436
InChI Key LSGBKABSSSIRJF-UHFFFAOYSA-N
Synonym 2,4,6-tris trifluoromethyl-1,3,5-triazine, s-triazine, 2,4,6-tris trifluoromethyl, 1,3,5-triazine, 2,4,6-tris trifluoromethyl, 2,4,6-tris trifluoromethyl-sym-triazine, 2,4,6-tris trifluoromethyl-s-triazine, tris trifluoromethyl-1,3,5-triazine, acmc-1cp6j, 5-26-01-00320 beilstein handbook reference, 1,3,5-triazine,2,4,6-tris trifluoromethyl, 2,4,6-tris trifluoromethyl-1,3,5 triazine
PubChem CID 9715
IUPAC Name 2,4,6-tris(trifluoromethyl)-1,3,5-triazine
SMILES FC(F)(F)C1=NC(=NC(=N1)C(F)(F)F)C(F)(F)F

Specifications

Density 1.596
Boiling Point 95°C to 96°C
Flash Point None
Refractive Index 1.322
Quantity 1 g
Beilstein 302339
Formula Weight 285.07
Percent Purity 98%
Chemical Name or Material 2,4,6-Tris(trifluoromethyl)-1,3,5-triazine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 206-709-3
RTECSNumber XZ2800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only