missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(2-pyridyl)-s-triazine, For Spectrophotometric Det. of Fe, 99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014058
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014058 1 g
111014060 5 g
111014059 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111014058 Supplier Merck Emd Millipore Supplier No. 932851G

2,4,6-Tris(2-pyridyl)-s-triazine is a Case and Koft compound generally used in determination of iron in a colorimetric analysis.

Reagent for the spectrophotometric determination of Fe(II) and of total Fe

Chemical Identifiers

CAS 3682-35-7
Molecular Formula C18H12N6
Molecular Weight (g/mol) 312.34
MDL Number MFCD00006045
InChI Key KMVWNDHKTPHDMT-UHFFFAOYSA-N
Synonym 2,4,6-Tri(2-pyridyl)-s-triazine; TPTZ
IUPAC Name tris(pyridin-2-yl)-1,3,5-triazine
SMILES C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 247°C to 249°C; 248°C to 251°C
Linear Formula C18H12N6
Quantity 1 g
Formula Weight 312.33
Percent Purity ≥99.0% (HPLC)
RTECSNumber XZ2050000