missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(2-pyridyl)-s-triazine, For Spectrophotometric Det. of Fe, 98%, MilliporeSigma™ Supelco™

Catalog No. 111014054
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111014054 1 g Glass Bottle
111014057 5 g Glass Bottle
111014056 10 g Glass Bottle
111014055 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111014054 Supplier Merck Emd Millipore Supplier No. T12531G

2,4,6-Tris(2-pyridyl)-s-triazine is a complexing reagent used in spectrophotometric analysis.

Chemical Identifiers

CAS 3682-35-7
Molecular Formula C18H12N6
Molecular Weight (g/mol) 312.34
MDL Number MFCD00006045
InChI Key KMVWNDHKTPHDMT-UHFFFAOYSA-N
Synonym 2,4,6-Tri(2-pyridyl)-s-triazine; TPTZ
IUPAC Name tris(pyridin-2-yl)-1,3,5-triazine
SMILES C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 247°C to 249°C
Packaging Glass Bottle
Linear Formula C18H12N6
Quantity 1 g
Formula Weight 312.33
Percent Purity ≥98%
Grade For spectrophotometric det. (of Fe)
RTECSNumber XZ2050000