missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triphenylaniline, 98%

CAS: 6864-20-6 | C24H19N | 321.423 g/mol

$176.18 - $1941.84

Chemical Identifiers

CAS 6864-20-6
Molecular Formula C24H19N
Molecular Weight (g/mol) 321.423
MDL Number MFCD00012371
InChI Key AGWKGKUGJDMKMT-UHFFFAOYSA-N
Synonym 2,4,6-triphenyl-anilin, 2,4,6-triphenylphenylamine, bidd:gt0713, 2,4,6-triphenylaniline, 3,5-diphenyl-1,1'-biphenyl-2-amine, 1,1':3',1-terphenyl-2'-amine,5'-phenyl, 5'-phenyl-1,1':3',1-terphenyl-2'-amine, 1,1':3',1-terphenyl-2'-amine, 5'-phenyl
PubChem CID 629425
IUPAC Name 2,4,6-triphenylaniline
SMILES C1=CC=C(C=C1)C2=CC(=C(C(=C2)C3=CC=CC=C3)N)C4=CC=CC=C4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2487103
View Documents
Thermo Scientific Chemicals
B2487103
1 g
Each for $176.18
Only null left
 
AAB2487106
View Documents
Thermo Scientific Chemicals
B2487106
5 g
Each for $545.48
Only null left
 
AAB2487114
View Documents
Thermo Scientific Chemicals
B2487114
25 g
Each for $1,941.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6864-20-6
321.423
AGWKGKUGJDMKMT-UHFFFAOYSA-N
629425
C1=CC=C(C=C1)C2=CC(=C(C(=C2)C3=CC=CC=C3)N)C4=CC=CC=C4
C24H19N
MFCD00012371
2,4,6-triphenyl-anilin, 2,4,6-triphenylphenylamine, bidd:gt0713, 2,4,6-triphenylaniline, 3,5-diphenyl-1,1'-biphenyl-2-amine, 1,1':3',1-terphenyl-2'-amine,5'-phenyl, 5'-phenyl-1,1':3',1-terphenyl-2'-amine, 1,1':3',1-terphenyl-2'-amine, 5'-phenyl
2,4,6-triphenylaniline

Specifications

6864-20-6
C24H19N
1 g
AGWKGKUGJDMKMT-UHFFFAOYSA-N
2,4,6-triphenylaniline
629425
98%
122°C to 125°C
MFCD00012371
2,4,6-triphenyl-anilin, 2,4,6-triphenylphenylamine, bidd:gt0713, 2,4,6-triphenylaniline, 3,5-diphenyl-1,1'-biphenyl-2-amine, 1,1':3',1-terphenyl-2'-amine,5'-phenyl, 5'-phenyl-1,1':3',1-terphenyl-2'-amine, 1,1':3',1-terphenyl-2'-amine, 5'-phenyl
C1=CC=C(C=C1)C2=CC(=C(C(=C2)C3=CC=CC=C3)N)C4=CC=CC=C4
321.423
321.43
2,4,6-Triphenylaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only