missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triphenylaniline, 98%

Catalog No. AAB2487103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2487103 1 g
AAB2487106 5 g
AAB2487114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2487103 Supplier Thermo Scientific Chemicals Supplier No. B2487103

CAS: 6864-20-6 | C24H19N | 321.423 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6864-20-6
Molecular Formula C24H19N
Molecular Weight (g/mol) 321.423
MDL Number MFCD00012371
InChI Key AGWKGKUGJDMKMT-UHFFFAOYSA-N
Synonym 2,4,6-triphenyl-anilin, 2,4,6-triphenylphenylamine, bidd:gt0713, 2,4,6-triphenylaniline, 3,5-diphenyl-1,1'-biphenyl-2-amine, 1,1':3',1-terphenyl-2'-amine,5'-phenyl, 5'-phenyl-1,1':3',1-terphenyl-2'-amine, 1,1':3',1-terphenyl-2'-amine, 5'-phenyl
PubChem CID 629425
IUPAC Name 2,4,6-triphenylaniline
SMILES C1=CC=C(C=C1)C2=CC(=C(C(=C2)C3=CC=CC=C3)N)C4=CC=CC=C4

Specifications

Melting Point 122°C to 125°C
Quantity 1 g
Formula Weight 321.43
Percent Purity 98%
Chemical Name or Material 2,4,6-Triphenylaniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only