missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triphenyl-s-triazine, 98+%

Catalog No. AC422190250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422190250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC422190250 Supplier Thermo Scientific Chemicals Supplier No. 422190250
Only null left

CAS: 493-77-6 | C21H15N3 | 309.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 493-77-6
Molecular Formula C21H15N3
Molecular Weight (g/mol) 309.37
MDL Number MFCD00006051
InChI Key HBQUOLGAXBYZGR-UHFFFAOYSA-N
Synonym cyaphenine, triphenyl-s-triazine, 1,3,5-triazine, 2,4,6-triphenyl, 2,4,6-triphenyl-s-triazine, kyaphenine, s-triphenyltriazine, 2,4,6-triphenyltriazine, triphenyl-1,3,5-triazine, s-triazine, 2,4,6-triphenyl-8ci, s-triazine, 2,4,6-triphenyl
PubChem CID 10305
IUPAC Name 2,4,6-triphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 231°C to 236°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 26, V,3, 26
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether,benzene,toluene,chcl3,soluble in acetic acid
Formula Weight 309.36
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4, 6-Triphenyl-s-triazine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-779-8
TSCA TSCA

RUO – Research Use Only