missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Triphenyl-1,3,5-triazine 98.0+%, TCI America™
$33.56 - $128.62
Chemical Identifiers
| CAS | 493-77-6 |
|---|---|
| Molecular Formula | C21H15N3 |
| Molecular Weight (g/mol) | 309.37 |
| MDL Number | MFCD00006051 |
| InChI Key | HBQUOLGAXBYZGR-UHFFFAOYSA-N |
| Synonym | cyaphenine, triphenyl-s-triazine, 1,3,5-triazine, 2,4,6-triphenyl, 2,4,6-triphenyl-s-triazine, kyaphenine, s-triphenyltriazine, 2,4,6-triphenyltriazine, triphenyl-1,3,5-triazine, s-triazine, 2,4,6-triphenyl-8ci, s-triazine, 2,4,6-triphenyl |
| PubChem CID | 10305 |
| IUPAC Name | triphenyl-1,3,5-triazine |
| SMILES | C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 493-77-6 | |
| 309.37 | |
| HBQUOLGAXBYZGR-UHFFFAOYSA-N | |
| 10305 | |
| C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1 |
| C21H15N3 | |
| MFCD00006051 | |
| cyaphenine, triphenyl-s-triazine, 1,3,5-triazine, 2,4,6-triphenyl, 2,4,6-triphenyl-s-triazine, kyaphenine, s-triphenyltriazine, 2,4,6-triphenyltriazine, triphenyl-1,3,5-triazine, s-triazine, 2,4,6-triphenyl-8ci, s-triazine, 2,4,6-triphenyl | |
| triphenyl-1,3,5-triazine |
Specifications
| 493-77-6 | |
| White-Yellow | |
| C21H15N3 | |
| 1 g | |
| HBQUOLGAXBYZGR-UHFFFAOYSA-N | |
| triphenyl-1,3,5-triazine | |
| 10305 | |
| ≥98.0% (GC) | |
| 2,4,6-Triphenyl-1,3,5-triazine |
| 236°C | |
| 230°C | |
| MFCD00006051 | |
| cyaphenine, triphenyl-s-triazine, 1,3,5-triazine, 2,4,6-triphenyl, 2,4,6-triphenyl-s-triazine, kyaphenine, s-triphenyltriazine, 2,4,6-triphenyltriazine, triphenyl-1,3,5-triazine, s-triazine, 2,4,6-triphenyl-8ci, s-triazine, 2,4,6-triphenyl | |
| C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 309.37 | |
| 309.37 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes