missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triphenyl-1,3,5-triazine 98.0+%, TCI America™

Catalog No. T27851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27851G 1 g
T27855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27851G Supplier TCI America Supplier No. T27851G

Chemical Identifiers

CAS 493-77-6
Molecular Formula C21H15N3
Molecular Weight (g/mol) 309.37
MDL Number MFCD00006051
InChI Key HBQUOLGAXBYZGR-UHFFFAOYSA-N
Synonym cyaphenine, triphenyl-s-triazine, 1,3,5-triazine, 2,4,6-triphenyl, 2,4,6-triphenyl-s-triazine, kyaphenine, s-triphenyltriazine, 2,4,6-triphenyltriazine, triphenyl-1,3,5-triazine, s-triazine, 2,4,6-triphenyl-8ci, s-triazine, 2,4,6-triphenyl
PubChem CID 10305
IUPAC Name triphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 236°C
Color White-Yellow
Boiling Point 230°C
Quantity 1 g
Formula Weight 309.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triphenyl-1,3,5-triazine