missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylphenyl isocyanate, 98%

Catalog No. p-7045962
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1261103 1 g
AAL1261106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1261103 Supplier Thermo Scientific Chemicals Supplier No. L1261103

CAS: 2958-62-5 | C10H11NO | 161.204 g/mol

2,4,6-Trimethylphenyl isocyanate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylphenyl isocyanate is used as an organic chemical synthesis intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines, bases and heat.

Chemical Identifiers

CAS 2958-62-5
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00019909
InChI Key HJHZRZFONUPQAA-UHFFFAOYSA-N
Synonym 2,4,6-trimethylphenyl isocyanate, mesityl isocyanate, 2,4,6-trimethylphenylisocyanate, acmc-1ckns, 2,4,6-trimethylbenzenisocyanate, 2-isocyanato-1,3,5-trimethyl-benzene, #
PubChem CID 595239
IUPAC Name 2-isocyanato-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)N=C=O)C

Specifications

Melting Point 43°C to 46°C
Boiling Point 218°C to 220°C
Flash Point 104°C (219°F)
Odor Irritating
Quantity 1 g
UN Number UN2811
Beilstein 2085601
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 161.21
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethylphenyl isocyanate
Show More Show Less
Hazard Category H300+H330-H315-H317-H319-H335
Hazard Statement GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only